Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncx-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269165.1 | NP_001256094.1 | 7 - 8 | 1466 | 125 | |
NM_001373058.1 | NP_001360010.1 | 8 - 9 | 1421 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |