Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072806.5 | NP_505207.1 | 9 - 10 | 1454 | 112 | |
GenBank mRNA | AY523511.1 | - | 9 - 10 | 1454 | 112 | |
X83887.1 | - | 1441 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |