Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dop-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028876.3 | NP_001024047.1 | 2 - 3 | 318 | 91 | |
NM_001269197.4 | NP_001256126.1 | 2 - 3 | 324 | 91 | ||
NM_001269198.4 | NP_001256127.1 | 2 - 3 | 319 | 91 | ||
NM_001373076.2 | NP_001360549.1 | 2 | 300 | 91 | ||
NM_001373077.2 | NP_001360548.1 | 2 | 300 | 91 | ||
NM_001373078.2 | NP_001360550.1 | 2 - 3 | 316 | 91 | ||
GenBank mRNA | AB125702.1 | - | 2 - 3 | 340 | 91 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Recommended Assay Caenorhabditis elegans Probe spans exons |