Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129369.6 | NP_001122841.1 | 4 - 5 | 655 | 82 | |
NM_073196.7 | NP_505597.2 | 4 - 5 | 637 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |