Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073557.6 | NP_505958.1 | 2 - 3 | 343 | 79 | |
GenBank mRNA | AF325178.1 | - | 2 - 3 | 331 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |