Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313082.3 | NP_001300011.1 | 8 - 9 | 1249 | 134 | |
NM_073559.8 | NP_505960.3 | 13 - 14 | 2271 | 134 | ||
NR_132532.1 | 12 - 13 | 1908 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |