Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269462.4 | NP_001256391.1 | 6 - 7 | 2007 | 67 | |
NM_001269463.3 | NP_001256392.1 | 6 - 7 | 1992 | 67 | ||
GenBank mRNA | AY275181.1 | - | 6 - 7 | 1986 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |