Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dct-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028706.7 | NP_001023877.1 | 9 - 10 | 1281 | 71 | |
NM_001028707.6 | NP_001023878.1 | 9 - 10 | 1287 | 71 | ||
NM_001028708.6 | NP_001023879.1 | 9 - 10 | 1289 | 71 | ||
NM_001028709.5 | NP_001023880.1 | 9 - 10 | 1289 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |