Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074058.4 | NP_506459.3 | 3 - 4 | 574 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |