Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276972.3 | NP_001263901.1 | 2 - 3 | 399 | 77 | |
NM_001276973.2 | NP_001263902.1 | 2 - 3 | 448 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |