Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074804.3 | NP_507205.2 | 8 - 9 | 1701 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |