Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dkf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269795.3 | NP_001256724.1 | 1 - 2 | 46 | 63 | |
NM_001269796.3 | NP_001256725.1 | 1 - 2 | 46 | 63 | ||
NM_074838.6 | NP_507239.2 | 1 - 2 | 109 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |