Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075082.7 | NP_507483.1 | 5 - 6 | 708 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |