Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001361781.3 | NP_001348781.1 | 1 - 2 | 301 | 88 | |
NM_075711.7 | NP_508112.1 | 1 - 2 | 301 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |