Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rme-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383525.1 | 1 - 2 | 119 | 79 | ||
NM_075807.5 | NP_508208.2 | 1 - 2 | 113 | 79 | ||
GenBank mRNA | AY750054.1 | - | 1 - 2 | 113 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |