Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029517.3 | NP_001024688.1 | 6 - 7 | 796 | 92 | |
NM_001029518.3 | NP_001024689.1 | 6 - 7 | 799 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |