Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313445.3 | NP_001300374.1 | 15 - 16 | 3538 | 113 | |
NM_001313446.4 | NP_001300375.1 | 17 - 18 | 3835 | 113 | ||
NM_001380960.1 | NP_001368582.1 | 17 - 18 | 3814 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |