Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lite-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076642.7 | NP_509043.3 | 4 - 5 | 509 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |