Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076728.4 | NP_509129.1 | 11 - 12 | 1222 | 72 | |
GenBank mRNA | Y08637.1 | - | 1222 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |