Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tyra-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029350.7 | NP_001024521.1 | 10 - 11 | 1425 | 73 | |
NM_001029351.6 | NP_001024522.1 | 10 - 11 | 1207 | 73 | ||
NM_001038448.5 | NP_001033537.1 | 10 - 11 | 1215 | 73 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |