Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gap-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129726.4 | NP_001123198.1 | 8 - 9 | 2127 | 65 | |
NM_001307914.4 | NP_001294843.1 | 12 - 13 | 2593 | 65 | ||
NM_001307916.4 | NP_001294845.1 | 17 - 18 | 3364 | 65 | ||
NM_001383623.1 | 15 - 16 | 2974 | 65 | |||
NM_077193.6 | NP_509594.3 | 17 - 18 | 3371 | 65 | ||
NM_077194.7 | NP_509595.1 | 18 - 19 | 3433 | 65 | ||
NM_077195.7 | NP_509596.1 | 16 - 17 | 4021 | 65 | ||
NM_077196.6 | NP_509597.1 | 15 - 16 | 3160 | 65 | ||
NM_077197.6 | NP_509598.1 | 14 - 15 | 2910 | 65 | ||
NM_077198.7 | NP_509599.1 | 12 - 13 | 3490 | 65 | ||
NM_171753.7 | NP_741873.1 | 15 - 16 | 3393 | 65 | ||
NM_171754.6 | NP_741874.1 | 8 - 9 | 2116 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |