Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077459.6 | NP_509860.1 | 4 - 5 | 543 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |