Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sto-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077542.7 | NP_509943.1 | 4 - 5 | 542 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |