Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lact-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077568.4 | NP_509969.2 | 14 - 15 | 2249 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |