Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313441.4 | NP_001300370.1 | 6 - 7 | 843 | 85 | |
NM_077861.6 | NP_510262.1 | 8 - 9 | 1306 | 85 | ||
GenBank mRNA | AF187013.1 | - | 8 - 9 | 1257 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |