Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srxa-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028460.2 | NP_001023631.1 | 3 - 4 | 517 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |