Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028745.1 | NP_001023916.1 | 4 - 5 | 482 | 186 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |