Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002999.1 | NP_001002999.1 | 593 | 102 | ||
GenBank mRNA | AB080972.1 | - | 593 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |