Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197044.1 | NP_001183973.1 | 1 - 2 | 179 | 105 | |
GenBank mRNA | AF049909.1 | - | 1 - 2 | 160 | 105 | |
HG965164.1 | - | 1 - 2 | 168 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Inventoried Probe spans exons |