Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003060.2 | NP_001003060.1 | 6 - 7 | 938 | 102 | |
GenBank mRNA | AF384054.1 | - | 5 - 6 | 894 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |