Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003001.1 | NP_001003001.1 | 2 - 3 | 82 | 102 | |
XM_022415467.1 | XP_022271175.1 | 2 - 3 | 149 | 102 | ||
GenBank mRNA | AF491301.1 | - | 2 - 3 | 82 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |