Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003225.1 | NP_001003225.1 | 2 - 3 | 167 | 111 | |
XM_005628346.3 | XP_005628403.1 | 5 - 6 | 480 | 111 | ||
XM_022427125.1 | XP_022282833.1 | 7 - 8 | 624 | 111 | ||
XM_022427126.1 | XP_022282834.1 | 9 - 10 | 749 | 111 | ||
XM_022427127.1 | XP_022282835.1 | 8 - 9 | 645 | 111 | ||
XM_022427128.1 | XP_022282836.1 | 9 - 10 | 699 | 111 | ||
XM_022427129.1 | XP_022282837.1 | 7 - 8 | 621 | 111 | ||
XM_022427130.1 | XP_022282838.1 | 6 - 7 | 579 | 111 | ||
XM_022427131.1 | XP_022282839.1 | 4 - 5 | 392 | 111 | ||
XM_022427132.1 | XP_022282840.1 | 4 - 5 | 389 | 111 | ||
XM_022427133.1 | XP_022282841.1 | 6 - 7 | 544 | 111 | ||
XM_022427134.1 | XP_022282842.1 | 6 - 7 | 541 | 111 | ||
XM_022427135.1 | XP_022282843.1 | 5 - 6 | 477 | 111 | ||
XM_022427136.1 | XP_022282844.1 | 5 - 6 | 486 | 111 | ||
XM_022427137.1 | XP_022282845.1 | 5 - 6 | 483 | 111 | ||
XM_022427138.1 | XP_022282846.1 | 3 - 4 | 347 | 111 | ||
XM_022427139.1 | XP_022282847.1 | 3 - 4 | 344 | 111 | ||
XM_022427140.1 | XP_022282848.1 | 2 - 3 | 160 | 111 | ||
GenBank mRNA | Z75156.1 | - | 2 - 3 | 167 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |