Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KITLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012735.2 | NP_001012753.1 | 2 - 3 | 323 | 110 | |
XM_022427652.1 | XP_022283360.1 | 2 - 3 | 334 | 110 | ||
GenBank mRNA | AY094361.1 | - | 1 - 2 | 82 | 110 | |
S53329.1 | - | 1 - 2 | 129 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |