Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANP32A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003013.2 | NP_001003013.2 | 6 - 7 | 686 | 74 | |
GenBank mRNA | AY162293.1 | - | 6 - 7 | 686 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |