Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012344.1 | NP_001012344.1 | 989 | 73 | ||
XM_005617078.3 | XP_005617135.1 | 1312 | 73 | |||
XM_005617079.3 | XP_005617136.1 | 1155 | 73 | |||
GenBank mRNA | AY875970.1 | - | 989 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |