Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-DRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014768.1 | NP_001014768.1 | 2 - 3 | 406 | 142 | |
GenBank mRNA | M29611.1 | - | 2 - 3 | 406 | 142 | |
M57529.1 | - | 2 - 3 | 355 | 142 | ||
M57535.1 | - | 2 - 3 | 355 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |