Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H3-3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_537232.4 | XP_537232.1 | 1 - 2 | 83 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |