Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014110202.2 | XP_013965677.2 | 2 - 3 | 392 | 112 | |
XM_535737.6 | XP_535737.5 | 2 - 3 | 382 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |