Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022420672.1 | XP_022276380.1 | 4 - 5 | 1543 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Canis familiaris Probe spans exons |