Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005641610.2 | XP_005641667.1 | 1 - 2 | 156 | 131 | |
XM_538121.5 | XP_538121.1 | 1 - 2 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |