Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022424466.1 | XP_022280174.1 | 13 - 14 | 1337 | 117 | |
XM_538386.6 | XP_538386.3 | 13 - 14 | 1199 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |