Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005633531.3 | XP_005633588.1 | 4 - 5 | 9124 | 61 | |
XM_005633532.3 | XP_005633589.1 | 4 - 5 | 815 | 61 | ||
XM_005633534.3 | XP_005633591.1 | 4 - 5 | 459 | 61 | ||
XM_022407716.1 | XP_022263424.1 | 4 - 5 | 9127 | 61 | ||
XM_542318.5 | XP_542318.3 | 4 - 5 | 605 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons |