Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAVIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_538754.5 | XP_538754.1 | 2 - 3 | 595 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons |