Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022422715.1 | XP_022278423.1 | 861 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris |