Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022415374.1 | XP_022271082.1 | 3 - 4 | 626 | 69 | |
XM_545759.6 | XP_545759.2 | 3 - 4 | 818 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |