Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CISD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624585.3 | XP_005624642.1 | 244 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris |