Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKDCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_849285.5 | XP_854378.1 | 2 - 3 | 801 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |