Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_535424.6 | XP_535424.1 | 5 - 6 | 1048 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |