Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX3X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005641254.3 | XP_005641311.1 | 8 - 9 | 889 | 56 | |
XM_005641255.2 | XP_005641312.1 | 8 - 9 | 892 | 56 | ||
XM_005641256.3 | XP_005641313.1 | 8 - 9 | 891 | 56 | ||
XM_856175.5 | XP_861268.1 | 8 - 9 | 887 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |