Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHFPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_858595.5 | XP_863688.1 | 4 | 776 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |